3-[1-(Dimethylamino)ethyl]phenol 97%

Leverantör: Apollo Scientific

Synonyms: 3-(1-(Dimethylamino)ethyl)phenol

OR962693-25G OR962693-5G
APOSOR962693-25GEA 7300 SEK
APOSOR962693-25G APOSOR962693-5G
3-[1-(Dimethylamino)ethyl]phenol 97%
3-[1-(Dimethylamino)ethyl]phenol
Formel: C₁₀H₁₅NO
MW: 165,236 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08272688
CAS nummer: 105601-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR