(4-Aminophenyl)(1-pyrrolidinyl)methanone 95%
Leverantör: Apollo Scientific
Synonyms: 4-(Pyrrolidinocarbonyl)aniline
OR962580-10G OR962580-1G OR962580-5G
APOSOR962580-10GEA 6775 SEK
APOSOR962580-10G APOSOR962580-1G APOSOR962580-5G
(4-Aminophenyl)(1-pyrrolidinyl)methanone 95%
(4-Aminophenyl)(1-pyrrolidinyl)methanone
Formel: C₁₁H₁₄N₂O
MW: 190,25 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02046794
CAS nummer: 56302-41-1