[2-(Pyrrolidin-1-ylcarbonyl)phenyl]amine 98%
Leverantör: Apollo Scientific
Synonyms:
2-(Pyrrolidin-1-ylcarbonyl)aniline
OR962460-10G
OR962460-1G
OR962460-5G
APOSOR962460-10GEA
10925
SEK
APOSOR962460-10G
APOSOR962460-1G
APOSOR962460-5G
[2-(Pyrrolidin-1-ylcarbonyl)phenyl]amine 98%
[2-(Pyrrolidin-1-ylcarbonyl)phenyl]amine
Formel:
C₁₁H₁₄N₂O Storage Temperature: Omgivande |
MDL Number:
MFCD00034732 CAS nummer: 52745-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...