3-(4-Methoxyphenoxy)propionic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-(4-Methoxyphenoxy)propanoic acid
OR962446-10G OR962446-1G OR962446-5G
APOSOR962446-10GEA 8925 SEK
APOSOR962446-10G APOSOR962446-1G APOSOR962446-5G
3-(4-Methoxyphenoxy)propionic acid 98%
3-(4-Methoxyphenoxy)propionic acid
Formel: C₁₀H₁₂O₄
MW: 196,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00020515
CAS nummer: 20811-60-3