4-Methyl-1-phenyl-2-pentanone 95%

Leverantör: Apollo Scientific

Synonyms: 4-Methyl-1-phenylpentan-2-one

OR961657-10G OR961657-1G
APOSOR961657-10GEA 2925 SEK
APOSOR961657-10G APOSOR961657-1G
4-Methyl-1-phenyl-2-pentanone 95%
4-Methyl-1-phenyl-2-pentanone
Formel: (C₆H₅)CH₂C(O)CH₂CH(CH₃)₂
MW: 176,26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00015639
CAS nummer: 5349-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR