N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 97%
Leverantör: Apollo Scientific
Synonyms: Fmoc-Cha-OH, N-Fmoc-3-cyclohexyl-L-alanine
OR960948-1G OR960948-25G OR960948-5G
APOSOR960948-1GEA 620 SEK
APOSOR960948-1G APOSOR960948-25G APOSOR960948-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-3-cyclohexyl-L-alanine
Formel: C₂₄H₂₇NO₄
MW: 393,48 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00065614
CAS nummer: 135673-97-1