2-(4-Methylthiazol-5-yl)ethylamine 97%
Leverantör: Apollo Scientific
OR960927-1G OR960927-5G
APOSOR960927-1GEA 2400 SEK
APOSOR960927-1G APOSOR960927-5G
2-(4-Methylthiazol-5-yl)ethylamine 97%
2-(4-Methylthiazol-5-yl)ethylamine
Formel: C₆H₁₀N₂S
MW: 142.22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08060722
CAS nummer: 58981-35-4