Methyl-2-methylthiazole-4-carboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Methyl 2-methylthiazole-4-carboxylate
OR960918-1G OR960918-25G OR960918-5G
APOSOR960918-1GEA 1475 SEK
APOSOR960918-1G APOSOR960918-25G APOSOR960918-5G
Methyl-2-methylthiazole-4-carboxylate 98%
Methyl-2-methylthiazole-4-carboxylate
Formel: C₆H₇NO₂S
MW: 157,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09870034
CAS nummer: 6436-60-8