1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde 98%
Leverantör: Apollo Scientific
Synonyms: 1-(Oxan-2-yl)pyrazole-5-carbaldehyde
OR960152-1G OR960152-5G
APOSOR960152-1GEA 1975 SEK
APOSOR960152-1G APOSOR960152-5G
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde 98%
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-carbaldehyde
Formel: C₉H₁₂N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD07186538
CAS nummer: 957483-88-4