Indol-3-ylacetic acid hydrazide
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-(1H-Indol-3-yl)acetohydrazide, Indole-3-acetic acid hydrazide, Indole-3-acetic hydrazide
OR9596-1G
OR9596-5G
APOSOR9596-1GEA
670
SEK
APOSOR9596-1G
APOSOR9596-5G
Indol-3-ylacetic acid hydrazide
Indol-3-ylacetic acid hydrazide
Formel:
C₁₀H₁₁N₃O MW: 189,22 g/mol Smältpunkt: 142…145 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00005634 CAS nummer: 5448-47-5 EINECS: 226-672-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...