4-sec-Butylphenol 95%
Leverantör: Apollo Scientific
OR957513-25G OR957513-100G OR957513-500G
APOSOR957513-25GEA 1300 SEK
APOSOR957513-25G APOSOR957513-100G APOSOR957513-500G
4-sec-Butylphenol 95%
4-sec-Butylphenol
Formel: C₁₀H₁₄O
MW: 150,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00002375
CAS nummer: 99-71-8