Methyl-4-cyanotetrahydro-2H-pyran-4-carboxylate 97%
Leverantör: Apollo Scientific
Synonyms: Methyl 4-cyanotetrahydro-2H-pyran-4-carboxylate
OR954127-1G OR954127-250MG
APOSOR954127-1GEA 6775 SEK
APOSOR954127-1G APOSOR954127-250MG
Methyl-4-cyanotetrahydro-2H-pyran-4-carboxylate 97%
Methyl-4-cyanotetrahydro-2H-pyran-4-carboxylate
Formel: C₈H₁₁NO₃
MW: 169,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11100237
CAS nummer: 362703-30-8