3-(3,5-Dimethyl-1H-pyrazol-1-yl)propan-1-amine 90%
Leverantör: Apollo Scientific
OR953714-1G OR953714-5G
APOSOR953714-1GEA 2250 SEK
APOSOR953714-1G APOSOR953714-5G
3-(3,5-Dimethyl-1H-pyrazol-1-yl)propan-1-amine 90%
3-(3,5-Dimethyl-1H-pyrazol-1-yl)propan-1-amine
Formel: C₈H₁₅N₃
Storage Temperature: Omgivande
MDL Number: MFCD00462177
CAS nummer: 62821-89-0