4-(3-Methylbutoxy)benzaldehyde 98%

Leverantör: Apollo Scientific

OR953653-1G OR953653-5G
APOSOR953653-1GEA 3875 SEK
APOSOR953653-1G APOSOR953653-5G
4-(3-Methylbutoxy)benzaldehyde 98%
4-(3-Methylbutoxy)benzaldehyde
Formel: C₁₂H₁₆O₂
MW: 192,26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02254125
CAS nummer: 18986-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR