3-Pyrazol-1-ylpropionic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-(1-Pyrazolyl)propionic Acid
OR953062-10G OR953062-1G OR953062-5G
APOSOR953062-10GEA 6800 SEK
APOSOR953062-10G APOSOR953062-1G APOSOR953062-5G
3-Pyrazol-1-ylpropionic acid 98%
3-Pyrazol-1-ylpropionic acid
Formel: C₆H₈N₂O₂
MW: 140,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01822327
CAS nummer: 89532-73-0