Methyl-3-nitrophenylacetate 97%
Leverantör: Apollo Scientific
Synonyms: Methyl 3-Nitrophenylacetate
OR953022-10G OR953022-1G OR953022-5G
APOSOR953022-10GEA 10025 SEK
APOSOR953022-10G APOSOR953022-1G APOSOR953022-5G
Methyl-3-nitrophenylacetate 97%
Methyl-3-nitrophenylacetate
Formel: C₉H₉NO₄
MW: 195,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08669939
CAS nummer: 10268-12-9