2-(4-Piperidyl)-2-propanol 96%
Leverantör: Apollo Scientific
Synonyms: 2-(4-Piperidyl)isopropyl Alcohol
OR953011-10G OR953011-25G OR953011-5G
APOSOR953011-10GEA 2150 SEK
APOSOR953011-10G APOSOR953011-25G APOSOR953011-5G
2-(4-Piperidyl)-2-propanol 96%
2-(4-Piperidyl)-2-propanol
Formel: C₈H₁₇NO
MW: 143,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06797159
CAS nummer: 22990-34-7