6-Chloroimidazo[1,2-b]pyridazin-3-amine 96%

Leverantör: Apollo Scientific

Synonyms: 3-Amino-6-chloroimidazo[1,2-b]pyridazine

OR952866-10G OR952866-1G OR952866-25G
APOSOR952866-10GEA 3700 SEK
APOSOR952866-10G APOSOR952866-1G APOSOR952866-25G
6-Chloroimidazo[1,2-b]pyridazin-3-amine 96%
6-Chloroimidazo[1,2-b]pyridazin-3-amine
Formel: C₆H₅ClN₄
MW: 168,5800 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09881451
CAS nummer: 166176-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR