4-Methoxy-3-nitropyridine

Leverantör: Apollo Scientific
Varning

OR9526-25G OR9526-5G
APOSOR9526-25GEA 1725 SEK
APOSOR9526-25G APOSOR9526-5G
4-Methoxy-3-nitropyridine
4-Methoxy-3-nitropyridine
Formel: C₆H₆N₂O₃
Smältpunkt: 72…77 °C
Storage Temperature: Omgivande
MDL Number: MFCD00209661
CAS nummer: 31872-62-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR