1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine 95%
Leverantör: Apollo Scientific
OR952572-100MG OR952572-1G OR952572-250MG
APOSOR952572-100MGEA 855 SEK
APOSOR952572-100MG APOSOR952572-1G APOSOR952572-250MG
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine 95%
1,2,3,4-Tetrahydro-1-methylpyrrolo[1,2-a]pyrazine
Formel: C₈H₁₂N₂
MW: 136,1900 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00296984
CAS nummer: 73627-18-6