5H-Pyrido[4,3-b]indole ≥98%
Leverantör: Apollo Scientific
Synonyms: gamma-Carboline
OR951880-1G OR951880-250MG
APOSOR951880-1GEA 1250 SEK
APOSOR951880-1G APOSOR951880-250MG
5H-Pyrido[4,3-b]indole ≥98%
5H-Pyrido[4,3-b]indole
Formel: C₁₁H₈N₂
MW: 168,20 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00965976
CAS nummer: 244-69-9