1-(4-Aminophenyl)pyrrolidin-2-one 98%
Leverantör: Apollo Scientific
Synonyms: 1-(4-Aminophenyl)-2-pyrrolidone
OR951681-10G OR951681-1G OR951681-5G
APOSOR951681-10GEA 9500 SEK
APOSOR951681-10G APOSOR951681-1G APOSOR951681-5G
1-(4-Aminophenyl)pyrrolidin-2-one 98%
1-(4-Aminophenyl)pyrrolidin-2-one
Formel: C₁₀H₁₂N₂O
MW: 176,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00100993
CAS nummer: 13691-22-0