2,4-Dichloro-6-methylpyrimidine-5-carbonitrile 95%
Leverantör: Apollo Scientific
OR951649-100MG OR951649-1G OR951649-250MG
APOSOR951649-100MGEA 1100 SEK
APOSOR951649-100MG APOSOR951649-1G APOSOR951649-250MG
2,4-Dichloro-6-methylpyrimidine-5-carbonitrile 95%
2,4-Dichloro-6-methylpyrimidine-5-carbonitrile
Formel: C₆H₃Cl₂N₃
MW: 188,0141 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12198893
CAS nummer: 56035-64-4