Ethyl imidazo[1,2-a]pyridine-3-carboxylate 97%
Leverantör: Apollo Scientific
OR951648-1G OR951648-250MG
APOSOR951648-1GEA 2575 SEK
APOSOR951648-1G APOSOR951648-250MG
Ethyl imidazo[1,2-a]pyridine-3-carboxylate 97%
Ethyl imidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₀H₁₀N₂O₂
MW: 190,2000 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11656766
CAS nummer: 123531-52-2