4-Cyano-4-phenylcyclohexanone 97%
Leverantör: Apollo Scientific
Synonyms: 4-Oxo-1-phenylcyclohexanecarbonitrile, 4-Cyano-4-phenylcyclohexanone
OR951015-1G OR951015-5G
APOSOR951015-1GEA 1625 SEK
APOSOR951015-1G APOSOR951015-5G
4-Cyano-4-phenylcyclohexanone 97%
4-Cyano-4-phenylcyclohexanone
Formel: C₁₃H₁₃NO
MW: 199,24 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00044816
CAS nummer: 25115-74-6