1-Ethyl-1H-pyrazole-5-carboxylic acid 98%

Leverantör: Apollo Scientific

OR950986-10G OR950986-1G OR950986-5G
APOSOR950986-10GEA 10275 SEK
APOSOR950986-10G APOSOR950986-1G APOSOR950986-5G
1-Ethyl-1H-pyrazole-5-carboxylic acid 98%
1-Ethyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₈N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD02090833
CAS nummer: 400755-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR