Ethyl-5-pyrimidinecarboxylate 98%
Leverantör: Apollo Scientific
Synonyms: Ethyl pyrimidine-5-carboxylate
OR950934-25G OR950934-5G
APOSOR950934-25GEA 3650 SEK
APOSOR950934-25G APOSOR950934-5G
Ethyl-5-pyrimidinecarboxylate 98%
Ethyl-5-pyrimidinecarboxylate
Formel: C₇H₈N₂O₂
MW: 152.153 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03094043
CAS nummer: 40929-50-8