4-(Ethoxymethyl)-2-methoxyphenol 95%

Leverantör: Apollo Scientific

Synonyms: Ethyl Vanillyl ether

OR950811-1G OR950811-5G
APOSOR950811-1GEA 635 SEK
APOSOR950811-1G APOSOR950811-5G
4-(Ethoxymethyl)-2-methoxyphenol 95%
4-(Ethoxymethyl)-2-methoxyphenol
Formel: C₁₀H₁₄O₃
MW: 182,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06409999
CAS nummer: 13184-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR