3-Ethyl-3-methylglutarimide 95%

Leverantör: Apollo Scientific

Synonyms: Bemegride, 4-Ethyl-4-methylpiperidine-2,6-dione

OR950148-10G OR950148-5G
APOSOR950148-10GEA 4050 SEK
APOSOR950148-10G APOSOR950148-5G
3-Ethyl-3-methylglutarimide 95%
3-Ethyl-3-methylglutarimide
Formel: C₈H₁₃NO₂
MW: 155,20 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006673
CAS nummer: 64-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR