3,4-Dimethyl-1H-pyrazole Phosphate 97%
Leverantör: Apollo Scientific
Synonyms: 3,4-Dimethyl-1H-pyrazole phosphate
OR949671-100G OR949671-25G OR949671-500G
APOSOR949671-100GEA 1025 SEK
APOSOR949671-100G APOSOR949671-25G APOSOR949671-500G
3,4-Dimethyl-1H-pyrazole Phosphate 97%
3,4-Dimethyl-1H-pyrazole Phosphate
Formel: C₅H₁₁N₂O₄P
MW: 194,13 g/mol
Smältpunkt: 167…170 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09842544
CAS nummer: 202842-98-6