Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 5-cyclopropyl-1H-pyrazole-3-carboxylate
OR945783-1G OR945783-5G
APOSOR945783-1GEA 3075 SEK
APOSOR945783-1G APOSOR945783-5G
Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Methyl-3-cyclopropyl-1H-pyrazole-5-carboxylate
Formel: C₈H₁₀N₂O₂
Storage Temperature: Omgivande
MDL Number: MFCD08559008
CAS nummer: 1036733-11-5