5-Chloromethyl-2-oxazolidinone 95%
Leverantör: Apollo Scientific
Synonyms: 5-(Chloromethyl)oxazolidin-2-one
OR944569-1G OR944569-25G OR944569-5G
APOSOR944569-1GEA 1300 SEK
APOSOR944569-1G APOSOR944569-25G APOSOR944569-5G
5-Chloromethyl-2-oxazolidinone 95%
5-Chloromethyl-2-oxazolidinone
Formel: C₄H₆ClNO₂
MW: 135,55 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00005269
CAS nummer: 22625-57-6