3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%

Leverantör: Apollo Scientific

Synonyms: (3-Nitro-5-(pyrrolidine-1-carbonyl)phenyl)boronic acid

OR9428-1G
APOSOR9428-1GEA 1525 SEK
APOSOR9428-1G
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid 98%
3-Nitro-5-(1-pyrrolidinylcarbonyl)phenylboronic acid
Formel: C₁₁H₁₃BN₂O₅
MW: 264.0400 g/mol
Smältpunkt: 181…183 °C
Storage Temperature: Kylskåp
MDL Number: MFCD07783873
CAS nummer: 871332-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR