4,5-Dimethyl-3-hydroxy-2(5H)-furanone 95%
Leverantör: Apollo Scientific
Synonyms: 3-Hydroxy-4,5-dimethylfuran-2(5H)-one, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one
OR941750-10G OR941750-25G OR941750-5G
APOSOR941750-10GEA 875 SEK
APOSOR941750-10G APOSOR941750-25G APOSOR941750-5G
4,5-Dimethyl-3-hydroxy-2(5H)-furanone 95%
4,5-Dimethyl-3-hydroxy-2(5H)-furanone
Formel: C₆H₈O₃
MW: 128,13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00059957
CAS nummer: 28664-35-9