Moc-Val-OH 95%

Leverantör: Apollo Scientific

Synonyms: N-(Methoxycarbonyl)-L-valine, MOC-Val-OH

OR940937-25G OR940937-100G
APOSOR940937-25GEA 650 SEK
APOSOR940937-25G APOSOR940937-100G
Moc-Val-OH 95%
Moc-Val-OH
Formel: C₇H₁₃NO₄
MW: 175,2 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12795357
CAS nummer: 74761-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR