2,5-Dimethoxybenzyl chloride 95%
Leverantör: Apollo Scientific
Synonyms: 2-(Chloromethyl)-1,4-dimethoxybenzene
OR940864-1G OR940864-5G
APOSOR940864-1GEA 1200 SEK
APOSOR940864-1G APOSOR940864-5G
2,5-Dimethoxybenzyl chloride 95%
2,5-Dimethoxybenzyl chloride
Formel: C₉H₁₁ClO₂
MW: 186,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00092195
CAS nummer: 3840-27-5