4-Amino-1-naphthonitrile 95%

Leverantör: Apollo Scientific

Synonyms: 4-Amino-1-naphthalenecarbonitrile

OR939997-1G OR939997-250MG
APOSOR939997-1GEA 2675 SEK
APOSOR939997-1G APOSOR939997-250MG
4-Amino-1-naphthonitrile 95%
4-Amino-1-naphthonitrile
Formel: C₁₁H₈N₂
MW: 168,1946 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00004022
CAS nummer: 58728-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR