7-Methoxy-2-naphthol 95%
Leverantör: Apollo Scientific
Synonyms: 7-Methoxynaphthalen-2-ol
OR939782-1G OR939782-25G
APOSOR939782-1GEA 0 SEK
APOSOR939782-1G APOSOR939782-25G
7-Methoxy-2-naphthol 95%
7-Methoxy-2-naphthol
Formel: C₁₁H₁₀O₂
MW: 174.2000 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00075494
CAS nummer: 5060-82-2