2'-Chloro-4'-nitroacetophenone 95%
Leverantör: Apollo Scientific
Synonyms:
1-(2-Chloro-4-nitrophenyl)ethanone
OR939665-1G
OR939665-250MG
APOSOR939665-1GEA
875
SEK
APOSOR939665-1G
APOSOR939665-250MG
2'-Chloro-4'-nitroacetophenone 95%
2'-Chloro-4'-nitroacetophenone
Formel:
C₈H₆ClNO₃ MW: 199,59 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00017340 CAS nummer: 67818-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...