Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%
Leverantör: Apollo Scientific
Synonyms: trans-4-Hydroxycyclohexanecarboxylic Acid Ethyl Ester, trans-Ethyl 4-hydroxycyclohexanecarboxylate
OR939383-10G OR939383-1G OR939383-25G
APOSOR939383-10GEA 4275 SEK
APOSOR939383-10G APOSOR939383-1G APOSOR939383-25G
Ethyl-trans-4-hydroxycyclohexanecarboxylate 98%
Ethyl-trans-4-hydroxycyclohexanecarboxylate
Formel: C₉H₁₆O₃
MW: 172,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16295187
CAS nummer: 3618-04-0