BrettPhos palladacycle 3rd generation
Leverantör: Apollo Scientific
Varning
Synonyms: [(2-Di-cyclohexylphosphino-3,6-dimethoxy-2′,4′,6′- triisopropyl-1,1′-biphenyl)-2-(2′-amino-1,1′ -biphenyl)]palladium(II) methanesulphonate, BrettPhos Pd G3
OR939181-1G OR939181-250MG OR939181-5G
APOSOR939181-1GEA 2325 SEK
APOSOR939181-1G APOSOR939181-250MG APOSOR939181-5G
BrettPhos palladacycle 3rd generation
BrettPhos palladacycle 3rd generation
Formel: C₄₈H₆₆NO₅PPdS
MW: 906,50 g/mol
Smältpunkt: 150…193 °C
Storage Temperature: Omgivande
MDL Number: MFCD22572666
CAS nummer: 1470372-59-8