2-(3-Bromopropyl)-1,3-dioxolane 95%
Leverantör: Apollo Scientific
Synonyms: 4-Bromobutyraldehyde Ethylene Acetal
OR939057-1G OR939057-250MG OR939057-5G
APOSOR939057-1GEA 2850 SEK
APOSOR939057-1G APOSOR939057-250MG APOSOR939057-5G
2-(3-Bromopropyl)-1,3-dioxolane 95%
2-(3-Bromopropyl)-1,3-dioxolane
Formel: C₆H₁₁BrO₂
MW: 195,06 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09868694
CAS nummer: 62563-07-9