Tetrahydrofuran-3-carbaldehyde 95%

Leverantör: Apollo Scientific

OR939036-100G OR939036-25G
APOSOR939036-100GEA 2800 SEK
APOSOR939036-100G APOSOR939036-25G
Tetrahydrofuran-3-carbaldehyde 95%
Tetrahydrofuran-3-carbaldehyde
Formel: C₅H₈O₂
MW: 100.12 g/mol
Kokpunkt: 186 °C
Smältpunkt: 33 °C
Densitet: 1.06 g/cm³ (25 °C)
Flampunkt: >87 °C
Storage Temperature: Kylskåp
MDL Number: MFCD02101976
CAS nummer: 79710-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR