(R)-(+)-N,α-Dimethylbenzylamine 95%

Leverantör: Apollo Scientific

Synonyms: (R)-N-Methyl-1-phenylethanamine, (R)-N-Methyl-alpha-phenylethylamine

OR938961-1G OR938961-5G
APOSOR938961-1GEA 1175 SEK
APOSOR938961-1G APOSOR938961-5G
(R)-(+)-N,α-Dimethylbenzylamine 95%
(R)-(+)-N,α-Dimethylbenzylamine
Formel: C₉H₁₃N
MW: 135,2062 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00044966
CAS nummer: 5933-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR