(3-Chlorophenyl)ethylamine 95%

Leverantör: Apollo Scientific

Synonyms: 3-Chloro-N-ethylaniline

OR938633-1G OR938633-250MG OR938633-500MG
APOSOR938633-1GEA 8225 SEK
APOSOR938633-1G APOSOR938633-250MG APOSOR938633-500MG
(3-Chlorophenyl)ethylamine 95%
(3-Chlorophenyl)ethylamine
Formel: C₈H₁₀ClN
MW: 155,63 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00028626
CAS nummer: 15258-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR