2,4-Hexanedione 97%
Leverantör: Apollo Scientific
OR938383-1G OR938383-5G OR938383-10G
APOSOR938383-1GEA 995 SEK
APOSOR938383-1G APOSOR938383-5G APOSOR938383-10G
2,4-Hexanedione 97%
2,4-Hexanedione
Formel: C₆H₁₀O₂
MW: 114,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00009318
CAS nummer: 3002-24-2