(3-Methoxyphenyl)acetyl chloride 95%
Leverantör: Apollo Scientific
Synonyms: 2-(3-Methoxyphenyl)acetyl chloride
OR938326-1G OR938326-25G OR938326-5G
APOSOR938326-1GEA 1550 SEK
APOSOR938326-1G APOSOR938326-25G APOSOR938326-5G
(3-Methoxyphenyl)acetyl chloride 95%
(3-Methoxyphenyl)acetyl chloride
Formel: C₉H₉ClO₂
MW: 184,6196 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00012244
CAS nummer: 6834-42-0