Propyl methanesulphonate 95%
Leverantör: Apollo Scientific
Synonyms:
Methanesulphonic acid Propyl Ester
OR938252-10G
OR938252-1G
OR938252-5G
APOSOR938252-10GEA
6350
SEK
APOSOR938252-10G
APOSOR938252-1G
APOSOR938252-5G
Propyl methanesulphonate 95%
Propyl methanesulphonate
Formel:
C₄H₁₀O₃S MW: 138,18 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00007561 CAS nummer: 1912-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...