cis-4-Hydroxycyclohexanecarboxylic acid 98%
Leverantör: Apollo Scientific
OR938037-10G OR938037-5G OR938037-25G
APOSOR938037-10GEA 1625 SEK
APOSOR938037-10G APOSOR938037-5G APOSOR938037-25G
cis-4-Hydroxycyclohexanecarboxylic acid 98%
cis-4-Hydroxycyclohexanecarboxylic acid
Formel: C₇H₁₂O₃
MW: 144,1700 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02093477
CAS nummer: 3685-22-1