1,4-Dichloro-2-butanol 95%
Leverantör: Apollo Scientific
OR937831-10G OR937831-1G OR937831-5G
APOSOR937831-10GEA 9000 SEK
APOSOR937831-10G APOSOR937831-1G APOSOR937831-5G
1,4-Dichloro-2-butanol 95%
1,4-Dichloro-2-butanol
Formel: C₄H₈Cl₂O
MW: 143,01 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00191343
CAS nummer: 2419-74-1